3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic acid 1,4:2,3-Dianhydride ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C3110-1G C3110-5G
TCIAC3110-1GEA 386 DKK
TCIAC3110-1G TCIAC3110-5G
3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic acid 1,4:2,3-Dianhydride ≥97.0% (by titrimetric analysis)
3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic acid 1,4:2,3-Dianhydride
Formel: C₁₀H₈O₆
MDL Number: MFCD22373052
CAS-nummer: 6053-46-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Methanolysis Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR