Cyclohexylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

C3101-1G C3101-5G
TCIAC3101-1GEA 247 DKK
TCIAC3101-1G TCIAC3101-5G
Cyclohexylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)
Cyclohexylboronic acid pinacol ester
Formel: C₁₂H₂₃BO₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04038749
CAS-nummer: 87100-15-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR