2-Chlor-4-methoxybenzoesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Chloro-p-anisic acid

C3077-1G C3077-5G
TCIAC3077-1GEA 222 DKK
TCIAC3077-1G TCIAC3077-5G
2-Chlor-4-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
2-Chlor-4-methoxybenzoesyre
Formel: C₈H₇ClO₃
Molekylvægt: 186.59 g/mol
MDL Number: MFCD00085943
CAS-nummer: 21971-21-1

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 210.0 to 214.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR