(Chloromethyl)trimethoxysilane ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C3073-5G C3073-25G
TCIAC3073-5GEA 378 DKK
TCIAC3073-5G TCIAC3073-25G
(Chloromethyl)trimethoxysilane ≥98.0% (by GC)
(Chloromethyl)trimethoxysilane
Formel: C₄H₁₁ClO₃Si
Kogepunkt: 156°C
Flammepunkt: –3°C
MDL Number: MFCD00468107
CAS-nummer: 5926-26-1
UN nummer: 1993
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR