3-Chlor-5-Fluoranilin ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C3052-1G
C3052-5G
TCIAC3052-1GEA
288
DKK
TCIAC3052-1G
TCIAC3052-5G
3-Chlor-5-Fluoranilin ≥97.0% (by GC, titration analysis)
3-Chlor-5-Fluoranilin
Formel:
C₆H₅ClFN Molekylvægt: 145.56 g/mol Kogepunkt: 98°C |
MDL Number:
MFCD03407962 CAS-nummer: 4863-91-6 |
Specifikationer Testresultater
Appearance | Orange to Amber to Dark red clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...