4-Chlor-2-Fluorphenylboronsyre (contains varying amounts of Anhydride)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C3048-1G
C3048-5G
TCIAC3048-1GEA
206
DKK
TCIAC3048-1G
TCIAC3048-5G
4-Chlor-2-Fluorphenylboronsyre (contains varying amounts of Anhydride)
4-Chlor-2-Fluorphenylboronsyre
Formel:
C₆H₅BClFO₂ Smeltepunkt: 250°C |
MDL Number:
MFCD02684293 CAS-nummer: 160591-91-3 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...