Cefmetazole sodium ≥92.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C3029-250MG
C3029-1G
TCIAC3029-250MGEA
419
DKK
TCIAC3029-250MG
TCIAC3029-1G
Cefmetazole sodium ≥92.0% (by HPLC)
Cefmetazole sodium
Formel:
C₁₅H₁₆N₇NaO₅S₃ |
MDL Number:
MFCD00214260 CAS-nummer: 56796-39-5 Merck Index: 14,01926 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 92.0 area% |
Purity (Ion exchange titration) | min. 97.0 % |
Specific rotation [a]20/D | +73.0 to +85.0 deg (C=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...