4H-Cyclopenta[2,1-b:3,4-b']dithiophene ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

C3021-200MG C3021-1G
TCIAC3021-200MGEA 468 DKK
TCIAC3021-200MG TCIAC3021-1G
4H-Cyclopenta[2,1-b:3,4-b']dithiophene ≥97.0% (by GC)
4H-Cyclopenta[2,1-b:3,4-b']dithiophene
Formel: C₉H₆S₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD20133900
CAS-nummer: 389-58-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR