Cyclopentanecarbaldehyde ≥90.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Stabilization: stabiliseret med hydroquinon

C3019-1ML C3019-5ML
TCIAC3019-1MLEA 271 DKK
TCIAC3019-1ML TCIAC3019-5ML
Cyclopentanecarbaldehyde ≥90.0% (by GC) stabilised
Cyclopentanecarbaldehyde
Formel: C₆H₁₀O
Kogepunkt: 40°C
Flammepunkt: 28°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00060798
CAS-nummer: 872-53-7
EINECS: 212-829-7
UN nummer: 1989
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR