Cy-vBRIDP® ≥90.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

C2981-200MG C2981-1G
TCIAC2981-200MGEA 189 DKK
TCIAC2981-200MG TCIAC2981-1G
Cy-vBRIDP® ≥90.0% (by HPLC)
Dicyclohexyl(1,1-diphenylprop-1-en-2-yl)phosphine
Formel: C₂₇H₃₅P
Smeltepunkt: 130°C
MDL Number: MFCD10565627
CAS-nummer: 384842-24-4

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 90.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR