2-(Chloromethyl)-4-methylquinazoline ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C2928-200MG C2928-1G
TCIAC2928-200MGEA 238 DKK
TCIAC2928-200MG TCIAC2928-1G
2-(Chloromethyl)-4-methylquinazoline ≥98.0% (by GC)
2-(Chloromethyl)-4-methylquinazoline
Formel: C₁₀H₉ClN₂
MDL Number: MFCD09807547
CAS-nummer: 109113-72-6
UN nummer: 3261
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR