4-Cyano-4''-n-pentyl-p-terphenyl ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C2910-1G
C2910-5G
TCIAC2910-1GEA
353
DKK
TCIAC2910-1G
TCIAC2910-5G
4-Cyano-4''-n-pentyl-p-terphenyl ≥98.0% (by GC)
4-Cyano-4''-n-pentyl-p-terphenyl
Formel:
C₂₄H₂₃N Smeltepunkt: 130°C |
MDL Number:
MFCD00799423 CAS-nummer: 54211-46-0 EINECS: 259-028-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Mesomorphic range | 128 to 240 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...