4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-hydroxypiperidine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol
C2909-5G
C2909-25G
TCIAC2909-5GEA
1090
DKK
TCIAC2909-5G
TCIAC2909-25G
4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-hydroxypiperidine ≥98.0% (by GC, titration analysis)
4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-hydroxypiperidine
Formel:
C₁₂H₁₃ClF₃NO |
MDL Number:
MFCD00066999 CAS-nummer: 21928-50-7 EINECS: 244-665-7 |
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...