2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C2908-1G
C2908-5G
TCIAC2908-1GEA
394
DKK
TCIAC2908-1G
TCIAC2908-5G
2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane ≥98.0% (by GC)
2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane
Formel:
C₁₂H₁₄ClFO₂ Kogepunkt: 112°C |
MDL Number:
MFCD00020909 CAS-nummer: 3308-94-9 EINECS: 221-993-9 |
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...