(3-Chloro-4-methoxyphenyl)methanamine hydrochloride ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Chloro-4-methoxybenzylamine hydrochloride

C2903-1G C2903-5G
TCIAC2903-1GEA 132 DKK
TCIAC2903-1G TCIAC2903-5G
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride ≥97.0% (by HPLC, titration analysis)
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride
Formel: C₈H₁₁Cl₂NO
MDL Number: MFCD01211460
CAS-nummer: 41965-95-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 250.0 to 254.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR