IR-813 p-Toluenesulphonate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: IR 813 tosylate, 2-Chloro-3-[2-(1,1,3-trimethylbenz[e]indolin-2-ylidene)ethylidene]-1-[2-(1,1,3-t, 2-[2-[2-Chloro-3-[2-(1,3-dihydro-1,1,3-trimethyl-2H-benz[e]indol-2-ylidene)ethylidene]-1-cyclohexen-1-yl]ethenyl]-1,1,3-trimethyl-1H-benz[e]indolium 4-Methylbenzenesulphonate, 2-(2-(2-Chloro-3-(2-(1,1,3-trimethyl-1H-benzo[e]indol-2(3H)-ylidene)ethylidene)cyclohex-1-en-1-yl)vinyl)-1,1,3-trimethyl-1H-benzo[e]indol-3-ium 4-methylbenzenesulphonate

C2886-1G C2886-5G
TCIAC2886-1GEA 411 DKK
TCIAC2886-1G TCIAC2886-5G
IR-813 p-Toluenesulphonate ≥98.0% (by HPLC, total nitrogen)
IR-813 p-Toluenesulphonate
Formel: C₄₇H₄₇ClN₂O₃S
Molekylvægt: 755.41 g/mol
MDL Number: MFCD03093252
CAS-nummer: 134127-48-3

Bestil nu

Specifikationer Testresultater

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 98.0 % (total of Ion-Pair)
Purity (with Total Nitrogen) min. 98.0 %
Lambda max. 810.0 to 816.0 nm (MeOH)
Absorbance (E1%1cm) min. 3000 (MeOH, 810.0 to 816.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR