Capecitabine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 5'-Deoxy-5-fluoro-N4-[(pentyloxy)carbonyl]cytidine, Pentyl (1-((2R,3R,4S,5R)-3,4-dihydroxy-5-methyltetrahydrofuran-2-yl)-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl)carbamate

C2878-1G C2878-5G
TCIAC2878-1GEA 476 DKK
TCIAC2878-1G TCIAC2878-5G
Capecitabine ≥98.0% (by HPLC, titration analysis)
Capecitabine
Formel: C₁₅H₂₂FN₃O₆
Molekylvægt: 359.35 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00930626
CAS-nummer: 154361-50-9
Merck Index: 14,01754

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +96.0 to +100.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR