N-(2-Cyanoethyl)-N-ethyl-m-toluidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-(N-Ethyl-m-toluidino)propionitrile, 3-[Ethyl(3-methylphenyl)amino]propionitrile

C2877-1G C2877-5G
TCIAC2877-1GEA 247 DKK
TCIAC2877-1G TCIAC2877-5G
N-(2-Cyanoethyl)-N-ethyl-m-toluidine ≥98.0% (by GC, titration analysis)
N-(2-Cyanoethyl)-N-ethyl-m-toluidine
Formel: C₁₂H₁₆N₂
Molekylvægt: 188.27 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00019859
CAS-nummer: 148-69-6
EINECS: 205-721-6

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR