N-Carbobenzoxy-L-serine benzyl ester ≥97.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N-Cbz-L-serine Benzyl Ester, Z-Ser-OBzl, Benzyl ((benzyloxy)carbonyl)-L-serinate
C2873-25G
C2873-5G
TCIAC2873-25GEA
1510
DKK
TCIAC2873-25G
TCIAC2873-5G
N-Carbobenzoxy-L-serine benzyl ester ≥97.0% (by HPLC, total nitrogen)
N-Carbobenzoxy-L-serine benzyl ester
Formel:
C₁₈H₁₉NO₅ |
MDL Number:
MFCD00037824 CAS-nummer: 21209-51-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 83.0 to 87.0 deg-C |
Specific rotation [a]20/D | +4.0 to +6.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...