2-Chloro-3-(chloromethyl)quinoline ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C2861-1G C2861-5G
TCIAC2861-1GEA 403 DKK
TCIAC2861-1G TCIAC2861-5G
2-Chloro-3-(chloromethyl)quinoline ≥98.0% (by GC)
2-Chloro-3-(chloromethyl)quinoline
Formel: C₁₀H₇Cl₂N
MDL Number: MFCD05865134
CAS-nummer: 90097-52-2
UN nummer: 3261
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR