2-Chloro-1-(4-fluorobenzyl)benzimidazole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Chloro-1-(4-fluorobenzyl)-1H-benzo[d]imidazole

C2850-5G C2850-25G
TCIAC2850-5GEA 214 DKK
TCIAC2850-5G TCIAC2850-25G
2-Chloro-1-(4-fluorobenzyl)benzimidazole ≥98.0% (by GC)
2-Chloro-1-(4-fluorobenzyl)benzimidazole
Formel: C₁₄H₁₀ClFN₂
Molekylvægt: 260.70 g/mol
MDL Number: MFCD00800240
CAS-nummer: 84946-20-3
EINECS: 284-624-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR