2-Cloro-4-nitrobenzamid ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Aklomide
C2829-5G
C2829-25G
TCIAC2829-5GEA
444
DKK
TCIAC2829-5G
TCIAC2829-25G
2-Cloro-4-nitrobenzamid ≥98.0% (by GC)
2-Cloro-4-nitrobenzamid
Formel:
C₇H₅ClN₂O₃ |
MDL Number:
MFCD00017119 CAS-nummer: 3011-89-0 EINECS: 221-143-7 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 170.0 to 174.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...