4-Chlor-3-(trifluormethyl)benzoesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Chloro-α,α,α-trifluoro-m-toluic acid

C2803-1G C2803-5G
TCIAC2803-1GEA 238 DKK
TCIAC2803-1G TCIAC2803-5G
4-Chlor-3-(trifluormethyl)benzoesyre ≥98.0% (by GC, titration analysis)
4-Chlor-3-(trifluormethyl)benzoesyre
Formel: C₈H₄ClF₃O₂
Molekylvægt: 224.57 g/mol
MDL Number: MFCD01631468
CAS-nummer: 1737-36-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 165.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR