(R)-(+)-4-Chloro-3-hydroxybutyronitrile ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C2763-1G
C2763-5G
TCIAC2763-1GEA
337
DKK
TCIAC2763-1G
TCIAC2763-5G
(R)-(+)-4-Chloro-3-hydroxybutyronitrile ≥96.0% (by GC)
(R)-(+)-4-Chloro-3-hydroxybutyronitrile
Formel:
C₄H₆ClNO |
MDL Number:
MFCD00273362 CAS-nummer: 84367-31-7 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Specific rotation [a]20/D | +12.0 to +14.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...