Chitin oligosaccharides (contains N-Acetylglucosamine)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C2762-25G
C2762-100G
TCIAC2762-25GEA
575
DKK
TCIAC2762-25G
TCIAC2762-100G
Chitin oligosaccharides (contains N-Acetylglucosamine)
Chitin oligosaccharides
CAS-nummer:
165039-45-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Total Nitrogen | 6.2 to 6.5 % |
Specific rotation [a]20/D | +17.0 to +23.0 deg (C=1、H2O) |
Water | max. 5.0 % |
Content of N-AG | max. 50.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...