4-Chlor-3-Fluorphenylboronsyre (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Chloro-3-fluorobenzeneboronic acid, (4-Chloro-3-fluorophenyl)metaboric acid, (4-Chloro-3-fluorophenyl)boronic acid, Dihydroxy(4-chloro-3-fluorophenyl)borane

C2749-1G C2749-5G
TCIAC2749-1GEA 214 DKK
TCIAC2749-1G TCIAC2749-5G
4-Chlor-3-Fluorphenylboronsyre (contains varying amounts of Anhydride)
4-Chlor-3-Fluorphenylboronsyre
Formel: C₆H₅BClFO₂
Smeltepunkt: 144°C
MDL Number: MFCD01319010
CAS-nummer: 137504-86-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR