3-(1-Cyanoethyl)benzoesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C2730-5G C2730-25G
TCIAC2730-5GEA 296 DKK
TCIAC2730-5G TCIAC2730-25G
3-(1-Cyanoethyl)benzoesyre ≥98.0% (by GC, titration analysis)
3-(1-Cyanoethyl)benzoesyre
Formel: C₁₀H₉NO₂
MDL Number: MFCD00002520
CAS-nummer: 5537-71-3
EINECS: 226-897-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR