6-Chloropyrazine-2-carboxylic acid ≥96.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

C2725-1G C2725-5G
TCIAC2725-1GEA 501 DKK
TCIAC2725-1G TCIAC2725-5G
6-Chloropyrazine-2-carboxylic acid ≥96.0% (by HPLC, titration analysis)
6-Chloropyrazine-2-carboxylic acid
Formel: C₅H₃ClN₂O₂
Molekylvægt: 158.54 g/mol
MDL Number: MFCD00837871
CAS-nummer: 23688-89-3

Bestil nu

Specifikationer Testresultater

Appearance White to Green to Brown powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR