1-Cyclopropyl-2-(2-fluorophenyl)ethanone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Cyclopropyl 2-Fluorobenzyl ketone

C2710-5G C2710-25G
TCIAC2710-5GEA 173 DKK
TCIAC2710-5G TCIAC2710-25G
1-Cyclopropyl-2-(2-fluorophenyl)ethanone ≥98.0% (by GC)
1-Cyclopropyl-2-(2-fluorophenyl)ethanone
Formel: C₁₁H₁₁FO
Kogepunkt: 61°C
MDL Number: MFCD10687164
CAS-nummer: 150322-73-9

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR