4-Chlor-1-Fluor-2-nitrobenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Chloro-2-fluoronitrobenzene
C2704-5G
C2704-25G
TCIAC2704-5GEA
509
DKK
TCIAC2704-5G
TCIAC2704-25G
4-Chlor-1-Fluor-2-nitrobenzen ≥98.0% (by GC)
4-Chlor-1-Fluor-2-nitrobenzen
Formel:
C₆H₃ClFNO₂ Molekylvægt: 175.55 g/mol Kogepunkt: 237°C Smeltepunkt: 10°C |
MDL Number:
MFCD00069418 CAS-nummer: 345-18-6 EINECS: 206-457-4 |
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...