2-(Chloromethyl)benzonitrile ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C2656-5G C2656-25G
TCIAC2656-5GEA 206 DKK
TCIAC2656-5G TCIAC2656-25G
2-(Chloromethyl)benzonitrile ≥98.0% (by GC)
2-(Chloromethyl)benzonitrile
Formel: C₈H₆ClN
Molekylvægt: 151.5929 g/mol
Kogepunkt: 252°C
MDL Number: MFCD00019745
CAS-nummer: 612-13-5
EINECS: 210-292-3
UN nummer: 2923
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR