2-Chloro-4-(dimethylamino)benzaldehyde ≥98.0% (by GC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C2606-1G C2606-5G
TCIAC2606-1GEA 337 DKK
TCIAC2606-1G TCIAC2606-5G
2-Chloro-4-(dimethylamino)benzaldehyde ≥98.0% (by GC, total nitrogen)
2-Chloro-4-(dimethylamino)benzaldehyde
Formel: C₉H₁₀ClNO
Molekylvægt: 183.64 g/mol
MDL Number: MFCD00021051
CAS-nummer: 1424-66-4
EINECS: 215-840-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR