2-Chlor-6-methyl-3-nitropyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C2594-1G
C2594-5G
C2594-25G
TCIAC2594-1GEA
238
DKK
TCIAC2594-1G
TCIAC2594-5G
TCIAC2594-25G
2-Chlor-6-methyl-3-nitropyridin ≥98.0% (by GC)
Formel:
C₆H₅ClN₂O₂ |
MDL Number:
MFCD03085820 CAS-nummer: 56057-19-3 |
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...