(S)-(+)-Clopidogrel sulphate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl (S)-2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetate sulphate, (S)-(+)-Clopidogrel hydrogensulphate, Methyl (S)-(+)-2-(2-Chlorophenyl)-2-[6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl]acetate sulphate

C2556-1G C2556-5G
TCIAC2556-1GEA 616 DKK
TCIAC2556-1G TCIAC2556-5G
(S)-(+)-Clopidogrel sulphate ≥98.0% (by HPLC, total nitrogen)
(S)-(+)-Clopidogrel sulphate
Formel: C₁₆H₁₆ClNO₂S
Molekylvægt: 419.89 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00876395
CAS-nummer: 120202-66-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical Isomer max. 0.5 %
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +54.0 to +58.0 deg (C=1, MeOH)

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR