N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl ((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)-L-valinate hydrochloride, N-(2'-Cyanobiphenyl-4-ylmethyl) L-valine methyl ester monohydrochloride

C2475-25G C2475-5G
TCIAC2475-25GEA 1510 DKK
TCIAC2475-25G TCIAC2475-5G
N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride ≥98.0% (by HPLC, titration analysis)
N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride
Formel: C₂₀H₂₂N₂O₂
MDL Number: MFCD08460094
CAS-nummer: 482577-59-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0fba51d7d9359d1bfb4e7e66a7fc08e072d9925&vid=d0fba51d7d9359d1bfb4e7e66a7fc08e072d9925&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +17.0 to +21.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR