3-Chloropropionaldehyde dimethyl acetal ≥93.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Chloro-3,3-dimethoxypropane

C2458-25G C2458-5G
TCIAC2458-25GEA 1470 DKK
TCIAC2458-25G TCIAC2458-5G
3-Chloropropionaldehyde dimethyl acetal ≥93.0% (by GC)
3-Chloropropionaldehyde dimethyl acetal
Formel: C₅H₁₁ClO₂
Molekylvægt: 138.59 g/mol
Kogepunkt: 45°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD16038653
CAS-nummer: 35502-06-8
UN nummer: 1993
ADR: 3 III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR