Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethylbenzylamine)palladium(II)] ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: SingaCycle(TM)-A1

C2372-1G C2372-200MG
TCIAC2372-1GEA 4350 DKK
TCIAC2372-1G TCIAC2372-200MG
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethylbenzylamine)palladium(II)] ≥97.0% (by titrimetric analysis)
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethylbenzylamine)palladium(II)]

This product is unavailable in the U.S.

Formel: C₃₆H₄₈ClN₃Pd
Molekylvægt: 664.67 g/mol
MDL Number: MFCD15072151
CAS-nummer: 930796-10-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR