Cyclohexyl-p-toluenesulphonate ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Cyclohexyl 4-methylbenzenesulphonate, Cyclohexyl Tosylate, p-Toluenesulphonic acid Cyclohexyl Ester

C2363-25G C2363-5G
TCIAC2363-25GEA 985 DKK
TCIAC2363-25G TCIAC2363-5G
Cyclohexyl-p-toluenesulphonate ≥98.0% (by HPLC)
Cyclohexyl-p-toluenesulphonate
Formel: C₁₃H₁₈O₃S
Molekylvægt: 254.34 g/mol
MDL Number: MFCD00014291
CAS-nummer: 953-91-3
EINECS: 213-468-8

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR