2-Chloroquinoline-3-carbaldehyde ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C2289-1G C2289-5G
TCIAC2289-1GEA 214 DKK
TCIAC2289-1G TCIAC2289-5G
2-Chloroquinoline-3-carbaldehyde ≥98.0% (by GC)
2-Chloroquinoline-3-carbaldehyde
Formel: C₁₀H₆ClNO
MDL Number: MFCD00130079
CAS-nummer: 73568-25-9

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 148.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR