Carbonylchlorobis(triphenylphosphine)iridium(I) ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Carbonylchloro bis(triphenylphosphine)iridium(I), VASKA'S COMPLEX, Chlorocarbonylbis(triphenylphosphine)iridium(I)

C2252-200MG C2252-1G
TCIAC2252-200MGEA 657 DKK
TCIAC2252-200MG TCIAC2252-1G
Carbonylchlorobis(triphenylphosphine)iridium(I) ≥95.0% (by titrimetric analysis)
Carbonylchlorobis(triphenylphosphine)iridium(I)
Formel: C₃₇H₃₀ClIrOP₂
MDL Number: MFCD00009590
CAS-nummer: 14871-41-1
EINECS: 238-941-6
UN nummer: 2811
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR