Cefazolin sodium salt ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Sodium (6R,7R)-7-(2-(1H-tetrazol-1-yl)acetamido)-3-(((5-methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
C2242-25G
C2242-5G
TCIAC2242-25GEA
1470
DKK
TCIAC2242-25G
TCIAC2242-5G
Cefazolin sodium salt ≥98.0% (by HPLC, titration analysis)
Cefazolin sodium salt
Formel:
C₁₄H₁₄N₈NaO₄S₃ Molekylvægt: 476.48 g/mol Smeltepunkt: 190°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00056883 CAS-nummer: 27164-46-1 EINECS: 248-278-4 Merck Index: 14,01917 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -15.0 to -24.0 deg (C=10, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...