2,2'-O-Cyclouridine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,2'-O-Anhydro-(1-beta-D-arabinofuranosyl)uracil, (2R,3R,3aS,9aR)-3-Hydroxy-2-(hydroxymethyl)-2,3,3a,9a-tetrahydro-6H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-6-one, 2,2'-Anhydrouridine

C2208-25G C2208-5G
TCIAC2208-25GEA 624 DKK
TCIAC2208-25G TCIAC2208-5G
2,2'-O-Cyclouridine ≥98.0% (by GC)
2,2'-O-Cyclouridine
Formel: C₉H₁₀N₂O₅
Smeltepunkt: 244°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00004945
CAS-nummer: 3736-77-4
EINECS: 223-107-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -17.0 to -23.0 deg (C=4, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR