N-Carbobenzoxy-D-valine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Cbz-D-Val-OH, Z-D-Val-OH, N-Cbz-D-valine

C2139-25G C2139-5G
TCIAC2139-25GEA 1230 DKK
TCIAC2139-25G TCIAC2139-5G
N-Carbobenzoxy-D-valine ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-D-valine
Formel: C₁₃H₁₇NO₄
MDL Number: MFCD00065703
CAS-nummer: 1685-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c94d1e58b6f40c82831df20969e29825756d8109&vid=c94d1e58b6f40c82831df20969e29825756d8109&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 58.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR