N-Carbobenzoxy-D-leucine dicyclohexylammonium salt ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N-Cbz-D-leucine Dicyclohexylammonium Salt, Z-D-Leu-OH.DCHA
C2135-1G
C2135-5G
TCIAC2135-1GEA
173
DKK
TCIAC2135-1G
TCIAC2135-5G
N-Carbobenzoxy-D-leucine dicyclohexylammonium salt ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-D-leucine dicyclohexylammonium salt
Formel:
C₁₄H₁₉NO₄ |
MDL Number:
MFCD10566885 CAS-nummer: 7662-58-0 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 152.0 to 156.0 deg-C |
Specific rotation [a]20/D | +7.0 to +8.0 deg (C=3, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...