Coenzyme Q10 ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (all-E)-2-(3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione, Ubiquinone 10, Ubidecarenone

C1971-1G C1971-100MG
TCIAC1971-1GEA 813 DKK
TCIAC1971-1G TCIAC1971-100MG
Coenzyme Q10 ≥98.0% (by HPLC)
Coenzyme Q10
Formel: C₅₉H₉₀O₄
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00042919
CAS-nummer: 303-98-0
EINECS: 206-147-9
Merck Index: 14,09843

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 47.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR