O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: O-(1H-6-Chlorobenzotriazol-1-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate, TCTU

C1926-25G C1926-5G
TCIAC1926-25GEA 542 DKK
TCIAC1926-25G TCIAC1926-5G
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥98.0% (by HPLC)
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate
Formel: C₁₁H₁₅BClF₄N₅O
Molekylvægt: 355.53 g/mol
MDL Number: MFCD04973270
CAS-nummer: 330641-16-2
UN nummer: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94d796593be307b58c25e883da327b9700c2886f&vid=572a3fe170641f5a954dcebfa3a39bbfaa8db22c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 19.3 to 19.9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR