kappa-Carrageenan
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C1804-25G
C1804-500G
TCIAC1804-25GEA
156
DKK
TCIAC1804-25G
TCIAC1804-500G
kappa-Carrageenan
kappa-Carrageenan
Formel:
C₁₂H₁₇O₁₃S |
MDL Number:
MFCD00151514 CAS-nummer: 11114-20-8 EINECS: 234-350-2 Merck Index: 14,01864 |
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Drying loss | max. 12.0 % |
Viscosity | 10.0 to 25.0 m Pa-s (0.3 %, H2O, 25 deg-C) |
Ash Content | max. 40.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...