(S)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C1796-5G C1796-1G
TCIAC1796-5GEA 493 DKK
TCIAC1796-5G TCIAC1796-1G
(S)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane ≥97.0% (by GC, titration analysis)
(S)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
Formel: C₆H₁₁ClO₂
Flammepunkt: 53°C
MDL Number: MFCD00273365
CAS-nummer: 60456-22-6
UN nummer: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a951919cea07044f31d5f0cbe01e3cb1ecae5261&vid=a951919cea07044f31d5f0cbe01e3cb1ecae5261&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Specific rotation [a]20/D -40.0 to -46.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR