4-(Chloromethyl)styrene ≥90.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
1-(Chloromethyl)-4-vinylbenzene, 4-Vinylbenzyl Chloride
Stabilization:
Stabilised with TBC + ONP + 2-nitro-p-cresol
C1792-25G
C1792-250G
TCIAC1792-25GEA
321
DKK
TCIAC1792-25G
TCIAC1792-250G
4-(Chloromethyl)styrene ≥90.0% (by GC) stabilised
4-(Chloromethyl)styrene
Formel:
C₉H₉Cl Kogepunkt: 229°C Flammepunkt: 103°C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00051362 CAS-nummer: 1592-20-7 EINECS: 216-471-2 UN nummer: 3265 ADR: 8,III |
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...