N-Carbobenzoxy-2-nitrobenzenesulphonamide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Cbz-2-nitrobenzenesulphonamide

C1757-25G C1757-5G
TCIAC1757-25GEA 3190 DKK
TCIAC1757-25G TCIAC1757-5G
N-Carbobenzoxy-2-nitrobenzenesulphonamide ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-2-nitrobenzenesulphonamide
Formel: C₁₄H₁₂N₂O₆S
MDL Number: MFCD06797080
CAS-nummer: 245365-64-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 143.0 to 147.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR